Learn More
Medchemexpress LLC 5'-Nucleotidase, Microorganism | 9027-73-0 | ≥50 U/mg protein | 200 U

Supplier: Medchemexpress LLC HYP3260200U
5′-Nucleotidase, Microorganism (CD73) is an intrinsic membrane glycoprotein acting as an ectoenzyme. It catalyzes the hydrolysis of 5-nucleotides into their respective nucleosides. This enzyme is defined by its ability to hydrolyze 1.0 μmole of inorganic phosphorus from adenosine 5′-monophosphate per minute at pH 9.0 and 37°C. It is soluble in water at concentrations of 10 mg/mL or greater and presents as a white to off-white solid. This product is strictly for research purposes and not intended for patient use.
- Acts as an ectoenzyme
- Catalyzes hydrolysis of 5-nucleotides
- Soluble in water at ≥ 10 mg/mL
- Presented as a white to off-white solid
- EC Number: 3.1.3.5
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.