Learn More
Medchemexpress LLC 5'-O-(4,4'-dimethoxytrityl)-2'-O-methyluridine | 103285-22-9 | 98.0% | 560.59 g/mol | C31H32N2O8 | 1 ML

Supplier: Medchemexpress LLC HYW02543810MM/1ML
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine is a protected 2'-O-methyluridine nucleoside used as a building block in oligonucleotide synthesis and nucleoside chemistry. It is supplied as a light yellow solid and as a 10 mM solution in DMSO for laboratory research applications.
- Protected 2'-O-methyl uridine analog for oligonucleotide synthesis.
- Available as a light yellow solid and as a 10 mM solution in DMSO.
- Purity 98.0% (data sheet).
- Molecular formula C31H32N2O8; molecular weight 560.59 g/mol.
- Supplied in multiple solid sizes for scale flexibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.