Learn More
Medchemexpress LLC 5'-O-(4,4'-dimethoxytrityl)-2'-O-methyluridine | 103285-22-9 | MFCD00274123 | 98.0% | 560.59 g/mol | C31H32N2O8 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02543810G
Encompass_Preferred
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine is a DMT-protected 2'-O-methyl uridine nucleoside used as a building block and intermediate for solid-phase oligonucleotide synthesis and for research on modified RNA. It is supplied as a high-purity solid or as a DMSO solution to facilitate incorporation into synthesis workflows.
  • DMT-protected 2'-O-methyl uridine suitable for oligonucleotide synthesis.
  • High purity (98.03%) for reliable reactions.
  • Available as powder and 10 mM solution in DMSO for flexibility.
  • Long-term storage stability when kept at recommended temperatures.
  • Datasheet, COA, and SDS available to support quality control.
Catalog No. 50-002-99691