Learn More
Medchemexpress LLC 5'-O-(4,4'-Dimethoxytrityl)-5-methoxyuridine | 2095417-73-3 | C31H32N2O9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1541675MG
Encompass_Preferred
5'-O-(4,4'-Dimethoxytrityl)-5-methoxyuridine is a purine nucleoside analog known for its broad antitumor activity, particularly against indolent lymphoid malignancies. Its mechanisms of action include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-76832