Learn More
Medchemexpress LLC 5'-O-DMT-N4-Bz-5-Me-dC | 104579-03-5 | 95.25% | 647.72 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13860110MM/1ML
Encompass_Preferred
5'-O-DMT-N4-Bz-5-Me-dC is a modified nucleoside that acts like nucleosides in DNA synthesis. It can be used in the synthesis of deoxyribonucleic acid or nucleic acid. These nucleoside analogues are also utilized in antiviral products to prevent viral replication in infected cells and have applications against various viruses such as hepatitis B, hepatitis C, herpes simplex, and HIV. Once phosphorylated, they function as antimetabolites by incorporating into growing DNA strands.
  • Modified nucleoside
  • Used in the synthesis of deoxyribonucleic acid or nucleic acid
  • Acts like nucleosides in DNA synthesis
  • Can be used in antiviral products
  • Effective against hepatitis B virus, hepatitis C virus, herpes simplex, and HIV
  • Works as an antimetabolite once phosphorylated
Catalog No. 50-003-50007