Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-O-(E)-p-coumaroylquinic acid | 5746-55-4 | 99.9% | 338.31 g/mol | C16H18O8 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN1054310MG

Encompass_Preferred

5-O-(E)-p-Coumaroylquinic acid is a quinic acid derivative reported to inhibit hepatitis B virus. Supplied as a high-purity research reagent in powder form, it is intended for in vitro antiviral and phytochemical studies and should be handled using standard laboratory safety procedures.

  • High purity (≈99.9%) powder for research use.
  • Reported antiviral activity against hepatitis B virus.
  • Molecular weight 338.31 g/mol; formula C16H18O8.
  • Supplied in milligram-scale packages suitable for small-scale experiments.
  • Storage: powder at -20°C for long-term storage, 4°C for short-term storage.

Catalog No. 50-002-95623


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.