Learn More
Medchemexpress LLC 5'-O-TBDMS-dA | 51549-30-5 | 99.9% | 365.50 | 1 ML

Supplier: Medchemexpress LLC HY13859910MM/1ML
5'-O-TBDMS-dA is a modified nucleoside that can be used to synthesize DNA or RNA. It is for research use only.
- Modified nucleoside
- Used to synthesize DNA or RNA
- Purity: 99.9%
- Molecular weight: 365.50
- Appearance: White to off-white solid
- Classified as oligonucleotide and nucleoside analog
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.