Learn More
Medchemexpress LLC 5'-O-TBDMS-dT | 40733-28-6 | 99.4% | 356.49 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY138597
Encompass_Preferred
5'-O-TBDMS-dT is a nucleoside with protective and modification effects, intended for research use only.
  • Involved in DNA/RNA synthesis pathway
  • Acts as a nucleoside antimetabolite or analog
  • Solid form storage at 4°C, protected from light
  • Solution form storage at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-48855