Learn More
Medchemexpress LLC 5'-ODMT cEt G Phosphoramidite (Amidite) | 945628-66-0 | 99.1% | 881.95 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY148504
Encompass_Preferred
5'-ODMT cEt G Phosphoramidite Amidite is a potent nucleic acid analog that demonstrates excellent safety and antisense activity. It is intended for research use only.
  • Enhances RNA-binding ability
  • Affects the construction of antisense oligonucleotides (ASOs)
  • Reduces ASO toxicity through alterations in nucleotide conformation and 5'-alkyl DNA modification
Catalog No. 50-003-63318