Learn More
Medchemexpress LLC 5-PAHSA 100mg | 1481636-41-2 | 538.89 g/mol | C34H66O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116193100MG
Encompass_Preferred
5-PAHSA (5-(palmitoyloxy)octadecanoic acid) is a fatty-acid-derived signaling lipid (a FAHFA) used as a research reagent and analytical standard for studies of lipid signaling, metabolism, inflammation, and insulin sensitivity. Molecular formula C34H66O4; molecular weight 538.89 g/mol. Intended for research use only.
  • Endogenous fatty acid ester of hydroxy fatty acid (FAHFA).
  • Reported anti-inflammatory and insulin-sensitizing activity in animal models.
  • Suitable as an analytical reference standard for LC-MS and biochemical assays.
  • Available as a solid in multiple pack sizes, including 100 mg.
  • Commonly used in metabolic and lipid signaling research workflows.
Catalog No. 50-002-29792