Learn More
Cayman Chemical 5-PAHSA-d31 100ug
SDP
Supplier:  Cayman Chemical 17197100 UG
Encompass_Preferred
An internal standard for the quantification of 5-PAHSA by GC- or LC-MS
Catalog No. 50-297-9789