Learn More
Medchemexpress LLC 5-Ph-IAA-AM | 2990123-10-7 | 99.0% | C19H17NO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14189410MM/1ML
Encompass_Preferred
5-Ph-IAA-AM is an eggshell-permeable analog of 5-Ph-IAA (HY-134653) that includes an acetoxymethyl (AM) group. This compound enhances protein degradation in laid embryos and can be utilized to reveal the roles of proteins in C. elegans, especially those involved in embryogenesis and development, by enabling temporally controlled protein degradation.
  • Enhanced protein degradation
  • Temporally controlled protein degradation
  • Eggshell-permeable
Catalog No. 50-003-54372