Learn More
Meso Scale Discovery U-PLEX CAR-T Cell Efficacy Combo 1, multiplex immunoassay, 8-plex, 5 plates, 96-well, sub- to low pg/mL sensitivity, for use with human samples, cell culture supernatant, plasma, serum
SDP
Supplier:  Meso Scale Discovery K15709K2
Encompass
U-PLEX CAR-T Cell Efficacy Combo 1, multiplex immunoassay, 8-plex, 5 plates, 96-well, sub- to low pg/mL sensitivity, for use with human samples, cell culture supernatant, plasma, serum
Catalog No. NC3234900
Add to Cart
Request a Quote