Learn More
Chemglass Life Sciences 5-Port, Vacuum Manifold
SDP
Supplier:  Chemglass Life Sciences CG444103
Encompass_Preferred
5-Port, Vacuum Manifold. Similar to the CG-4440 vacuum manifold, but with BOTH ENDS OF INERT GAS LINE OPEN to permit connection to the gas source and an oil bubbler. Hose connections have a 10mm small serration and a 11mm large serration.
Catalog No. 50-873-347