Learn More
Medchemexpress LLC 5α-Pregnane-3β,6α-diol-20-one | 21853-11-2 | ≥98.0% | 334.49 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY109564100MG
Encompass_Preferred
5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone that can be produced in starved androgen-responsive prostate cancer cells. It is formed from progesterone in human fibroblasts and from pregnenolone and progesterone in C4.2 cell lines, with nearly complete conversions. Its formation is independent of CYP17A1 and relies on 3β-HSD activity.
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite
  • For research use only
Catalog No. 50-003-43259