Learn More
Medchemexpress LLC 5α-Pregnane-3β,6α-diol-20-one | 21853-11-2 | ≥98.0% | 334.49 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10956450MG
Encompass_Preferred
5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone that can be produced in starved androgen-responsive prostate cancer cells. It is formed from progesterone in human fibroblasts and from early steroid precursors in C4.2 cell lines, independently of CYP17A1 but relying on 3β-HSD activity.
  • Mitogenic metabolite of progesterone.
  • Can be produced in starved androgen-responsive prostate cancer cells.
  • Formed from progesterone in human fibroblasts.
  • Formed from early steroid precursors in C4.2 cell lines.
  • Formation is independent of CYP17A1.
  • Formation relies on 3β-HSD activity.
Catalog No. 50-003-43116