Learn More
Medchemexpress LLC 5-Pyrimidylboronic acid | 109299-78-7 | MFCD03002366 | 99.9% | C4H5BN2O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY592265G
Encompass_Preferred
5-Pyrimidylboronic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. It facilitates analogous one-pot copper-catalyzed DBAD coupling and condensation, and its utility in Suzuki-Miyaura coupling enables the efficient coupling of boronic acids with carbon halides and surrogates. This product is intended for research applications only.
  • Biochemical reagent for life science research
  • Supports analogous one-pot copper-catalyzed DBAD coupling and condensation
  • Enables Suzuki-Miyaura coupling for boronic acids with carbon halides and surrogates
Catalog No. 50-004-34214