Learn More
Medchemexpress LLC HY-76948 5mg Medchemexpress, 5-R-Rivaroxaban CAS:865479-71-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY769485MG
Encompass_Preferred
Medchemexpress, HY-76948 5mg 5-R-Rivaroxaban CAS:865479-71-6 5-R-Rivaroxaban is (R)-enantiomer of Rivaroxaban. Rivaroxaban (BAY 59-7939) is a highly potent and selective, direct Factor Xa (FXa) inhibitor, achieving a strong gain in anti-FXa potency (IC50 0.7 nM; Ki 0.4 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2283