Learn More
MSC E.R. Wagner Rigid Top Plate Caster, Polyurethane, 5 Inch Wheel Diameter, 450 Pound Capacity
Supplier: MSC 57078032
This rigid top plate caster features a durable polyurethane wheel, providing reliable performance for various applications. With a substantial load capacity and precise dimensions, it is suitable for medium-duty tasks requiring robust mobility.
- Wheel diameter of 5 inches.
- Constructed with polyurethane wheel material.
- Features a rigid top plate mount.
- Equipped with a roller bearing for smooth operation.
- Wheel width measures 1-1/2 inches.
- Overall height is 6-3/8 inches.
- Capable of handling a load up to 450 pounds.
- Ideal for medium-duty applications.
- Finished with a protective zinc coating.