Learn More
Apexbio Technology LLC 5-ROX 216699-35-3 5mg
SDP
Supplier:  Apexbio Technology LLC B82125
Encompass_Preferred
5-ROX (216699-35-3) is a fluorescent dye within the rhodamine family 5-ROX contains a carboxyl functional group enabling covalent labeling of biomolecules such as nucleic acids proteins and oligonucleotides Based on its optical properties including excitation at 580 nm and emission at 604 nm 5-ROX facilitates visualization and detection in analytical assays It is routinely employed for labeling applications in quantitative PCR (qPCR) probes fluorescence in situ hybridization (FISH) and flow cytometry-based biomolecular analysis Based on these characteristics 5-ROX holds research potential in biomolecular localization quantitation and interaction studies within biomedical research
Catalog No. 50-000-91499