Learn More
Medchemexpress LLC 5-(tert-Butyl)isobenzofuran-1,3-dione | 32703-79-0 | 96.0% | 204.23 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW012482
Encompass_Preferred
5-(tert-Butyl)isobenzofuran-1,3-dione, also known as 4-tert-Butylphthalic Anhydride, is an intermediate and dye. It is primarily used for research purposes in laboratory chemical manufacturing and has not been validated for medical applications.
  • Intermediate and dye
  • Suitable for research use
  • Used in laboratory chemical manufacturing
  • Functions as a fluorescent dye
  • Applicable in labeling chemistry
  • Utilized in chemical stain analysis
Catalog No. 50-004-52435