Learn More
Medchemexpress LLC 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[(1Z)-2-(4-methyl-5-thiazolyl)ethenyl]-8-oxo-, (2,2-dimethyl-1-oxopropoxy)methyl ester, (6R,7R)- | 117467-28-4 | 99.29% | 620.72 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17452A100MG
Encompass_Preferred
Cefditoren Pivoxil (ME 1207) is a broad-spectrum, third-generation, oral cephalosporin antibacterial. It shows enhanced stability against many common β lactamases and is active against both Gram-negative and Gram-positive organisms. This product is suitable for research into infectious diseases, including acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
  • Broad-spectrum antibacterial activity
  • Third-generation oral cephalosporin
  • Enhanced stability against common β lactamases
  • Active against Gram-negative organisms
  • Active against Gram-positive organisms
  • Useful in research for various infection diseases
Catalog No. 50-003-86633