Learn More
Medchemexpress LLC 5-(thieno[2,3-d]pyrimidin-4-ylamino)naphthalen-1-ol | 852678-17-2 | 99.0% | C16H11N3OS | 10MG

Supplier: Medchemexpress LLC HY14702610MG
CDK9-IN-15 is a small-molecule research compound described as a potent cyclin-dependent kinase 9 (CDK9) inhibitor, suitable for biochemical and cellular studies investigating CDK9-related pathways. It is supplied as a solid and intended for laboratory research use only.
- Potent CDK9 inhibition suitable for biochemical and cellular assays.
- High purity (98.9%) supporting reproducible experimental results.
- Molecular formula C16H11N3OS; molecular weight 293.34 g·mol-1.
- Solid form, brown to black; typically soluble in DMSO for assay preparation.
- Recommended storage: 4°C protected from light; in solvent store at -80°C for long-term.
- Available in small milligram pack sizes appropriate for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.