Learn More
Medchemexpress LLC 5-(trifluoromethyl)uracil | 54-20-6 | MFCD00006024 | 99.6% | 180.09 g/mol | C5H3F3N2O2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01860310G
Encompass_Preferred
5-(Trifluoromethyl)uracil is a fluorinated uracil derivative supplied as a solid reagent for life-science research and synthetic chemistry. It is used as a building block and research tool in nucleic acid chemistry and related studies. CAS 54-20-6; molecular formula C5H3F3N2O2; molecular weight ~180.09 g/mol.
  • High purity suitable for research applications.
  • Solid form simplifies storage and handling.
  • Useful as a building block for nucleic acid analogue synthesis.
  • Characterized by CAS number and molecular formula for traceability.
  • Available in multiple package sizes to support different workflows.
Catalog No. 50-003-00847