Learn More
Cayman Chemical 5 Z 8 Z 14 ZEicosatrienoic Ac
SDP
Supplier:  Cayman Chemical 10009733500 UG
Encompass_Preferred
5(Z),8(Z),14(Z)-Eicosatrienoic acid is a PUFA that can be a substrate for 5-LO. 5-LO from RBL-1 cells converts this fatty acid to 5-hydroxy-6,8,14-eicosatrienoic acid and 5-hydroperoxy-6,8,14-eicosatrienoic acid. Due to the lack of a double bond at C-11, this particular fatty acid cannot be used in LTA formation.{14955}
Catalog No. 50-288-7781