Learn More
Bellco Glass, Inc. 50-200µL Pipette, MLA, Digital, Red
SDP
Supplier:  Bellco Glass, Inc. 451900078
Encompass_Preferred
50-200µL Pipette, MLA, Digital, RedTHE DIAL-IN ADJUSTABLE MODELPrecise ‘click stop’ volume adjustmentSingle-stroke “to deliver” operationBuilt in tip-ejectorAvailable in 3 volumes; 10-50 μL, 50-200 μL, 200-1000 μLFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume50-200μLVistaLab Tip Fit SizeSmall 250μLUniversal FitNoAccuracy±1% at 200μL | ±1% at 125μL | ±2% at 50μLPrecision0.50% at 200μL | 0.43% at 125μL | 0.80% at 50μL
Catalog No. 50-233-0040