Learn More
Pacer Global LLC 50 ml Reservoir, White, Sterile, 5 per pack, 200 per case
SDP
Supplier:  Pacer Global LLC MLSMRWS50/DEL
Encompass_Preferred
50mL Disposable Reagent Reservoir. Sterile. Polystyrene. White. Made from virgin grade resin. Lot Certified RNase/DNase-free. Non-Pyrogenic, Animal Free, Human DNA Free. Designed to accommodate single & multichannel pipettors. Stacking lugs prevent crushing & facilitate separation. Graduation marks on the interior walls. Pour-off spouts in each corner. 5/pack, 200/case.
Catalog No. NC1865282