Learn More
Chemglass Life Sciences Chemglass 50 μL Automation Clear Tips for Tecan, Sterile - 96 x 2 blister packs (2,304/case)
SDP
Supplier:  Chemglass Life Sciences CLS3499A101
Encompass_Preferred
50 μL sterile automation pipette tips designed for use with automated liquid-handling systems. Made from medical-grade polypropylene and produced in a controlled clean-room environment, the tips are DNase/RNase free and electron-beam sterilized for reliable performance in high-throughput workflows.
  • Sterile, electron beam sterilized.
  • DNase/RNase free for molecular biology applications.
  • Medical-grade polypropylene construction.
  • Packaged in blister packs to support automation loading.
  • Produced in a 100,008 grade clean room for controlled purity.
  • Packaged as 96 x 2 blister packs; 2,304 tips per case for bulk workflows.
Catalog No. 50-273-3247