Learn More
Chemglass Life Sciences 50 μL Automation Clear Pipette Tips for Hamilton Systems, Sterile, Boxed, With Barcode - 96/rack, 10 racks/pack
SDP
Supplier:  Chemglass Life Sciences CLS3499A062
Encompass_Preferred
These 50 μL automation clear pipette tips are sterile, barcode-identified, and box-packed for automated liquid-handling workflows. Made from medical-grade polypropylene and produced in a clean-room environment, the tips include aerosol-resistant filters and are DNase/RNase-free with electron-beam sterilization to reduce contamination risk.
  • 50 μL nominal volume for precise low-volume transfers.
  • Box-packed racks: 96 tips per rack, 10 racks per pack, 50 racks per case.
  • Sterile, electron-beam sterilized to ensure low bioburden.
  • Medical-grade polypropylene construction.
  • DNase/RNase-free and produced in a clean-room environment.
  • Aerosol-resistant filter to reduce sample contamination.
  • Barcode on racks for inventory and automation tracking.
Catalog No. 50-273-3259