Learn More
Chemglass Life Sciences 50 μL Automation Clear Pipette Tips for Hamilton systems, sterile, box-packed, 96 tips/rack
SDP
Supplier:  Chemglass Life Sciences CLS3499A061
Encompass_Preferred
50 μL automation clear pipette tips designed for high-throughput automated liquid handling. These sterile, box-packed tips include rack barcodes for tracking and are supplied in automation-ready racks for reliable, low-retention pipetting in genomics, proteomics, and other laboratory workflows. Compatible with Hamilton automation systems.
  • 50 μL volume
  • Sterile and electron beam sterilized
  • Medical-grade polypropylene construction
  • DNase/RNase free
  • Barcode on racks for identification and tracking
  • Packaged 96 tips per rack; 10 racks per pack; 50 racks per case
  • Produced in a 100,049 grade clean room
Catalog No. 50-273-3260