Learn More
Chemglass Life Sciences Chemglass 50 μL automation clear pipette tips for Tecan systems, sterile, 96/rack (10 racks/pk)
SDP
Supplier:  Chemglass Life Sciences CLS3499A401
Encompass_Preferred
These 50 μL automation clear pipette tips are designed for high-throughput automated liquid handling systems and are supplied sterile in rack packaging to support reproducible pipetting in genomics, proteomics, immunoassays, metabolomics, and biopharmaceutical workflows. Compatible with Tecan automated liquid handling systems.
  • 50 μL nominal volume.
  • Medical-grade polypropylene.
  • Sterile; electron-beam sterilized.
  • DNase/RNase free.
  • Produced in 100,008 grade clean room.
  • Packaging: 96 tips per rack; 10 racks per pack; 50 racks per case.
Catalog No. 50-273-3237