Learn More
Chemglass Life Sciences 50 μL Automation Conductive Filter Tips for Hamilton Systems, Sterile, Barcode-Enabled - 96/Pack, 2,304/Case
SDP
Supplier:  Chemglass Life Sciences CLS3499A011
Encompass_Preferred
50 μL automation conductive filter tips designed for automated Hamilton liquid handling systems. Sterile and barcode-enabled for lot traceability; constructed from medical-grade polypropylene with aerosol-resistant conductive filters to protect samples and support reliable conductivity detection in high-throughput workflows.
  • 50 μL tip volume.
  • Sterile, electron beam sterilized.
  • 96 tips per pack; 2,304 tips per case.
  • Barcode enabled for traceability.
  • Medical-grade polypropylene construction.
  • DNase/RNase free.
  • Aerosol-resistant conductive filter for contamination control.
  • Produced in a 100,049 grade clean room.
Catalog No. 50-273-3281