Learn More
Chemglass Life Sciences Automation Conductive Filter Tips for Hamilton System, 50 μL, Sterile, Boxed, With Barcode
SDP
Supplier:  Chemglass Life Sciences CLS3499A031
Encompass_Preferred
50 μL automation conductive filter tips designed for Hamilton liquid-handling systems. Packaged sterile in boxed nests with barcode for rack tracking, these tips include an internal filter and conductive properties for reliable automated dispensing.
  • Volume: 50 μl
  • Pack format: 96 per rack; 10 racks per pack; 50 racks per case
  • Compatibility: Hamilton automated systems
  • Material: medical-grade polypropylene
  • Features: conductive, filter, sterile
  • Barcode: integrated for rack tracking
Catalog No. 50-273-3272