Learn More
Molecular Dimensions 50% V/V PEG SMEAR HIGH (250ML)
SDP
Supplier:  Molecular Dimensions MD2250260
Encompass
PEG is an effective crystallization agent due to its protein-protein interaction promoting properties. The BCS Screen overcomes the impracticality of sampling all PEG varieties that has led to a few variants predominating in the most common screens. PEG Smears are designed primarily for efficient screening of chemical space. It is generally thought that well-defined, single PEG species produce the best diffracting crystals. Thus, optimization might involve identifying the single PEG compound that produces the best crystal for structural studies. MOLECULAR DIMENSIONS INC mfg# MD2-250-260
Catalog No. NC2064002