Learn More
Bellco Glass, Inc. 500 mL PC Conical Erlenmeyer Flasks, Flat Bottom, Vent Filter Caps, Sterile,1/pk, 12/cs
SDP
Supplier:  Bellco Glass, Inc. 451400192
Encompass_Preferred
500 mL PC Conical Erlenmeyer Flasks, Flat Bottom, Vent Filter Caps, Sterile,1/pk, 12/cs. Ideal for shaker culture applications. There are two kinds of bottle materials, both PETG and PC are available. PC bottle or PEFG bottleSterilized by E-BeamNonpyrogenicIndividually wrapped in a plastic bag.Both vent caps and plug seal caps are available.Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.
Catalog No. 50-233-0134