Learn More
Medchemexpress LLC Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate | 28705-46-6 | MFCD00227484 | 99.8% | 289.33 | C16H19NO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW08743310MM/1ML
Encompass_Preferred
Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate is a small-molecule inhibitor of human monoamine oxidase B (hMAO-B) with an IC50 of 45.52 μM. It is supplied as a 10 mM solution in DMSO and in multiple solid sizes, and is provided with supporting documentation for quality and handling.
  • Potent inhibitor of hMAO-B, IC50 = 45.52 μM.
  • Available as 10 mM 1 mL solution in DMSO and as solids from 250 mg to 50 g.
  • High purity, 99.78%.
  • Characterized by CAS number 28705-46-6, formula C16H19NO4, molecular weight 289.33.
  • Supplied with datasheet, certificate of analysis, and SDS for quality and safety information.
Catalog No. 50-002-08411