Learn More
Medchemexpress LLC MMAF-d8 hydrochloride | 1415246-68-2 | 99.7% | 776.47 | C39H58D8ClN5O8 | 10 MG

Supplier: Medchemexpress LLC HY15579AS10MG
MMAF-d8 hydrochloride is the deuterated hydrochloride salt of monomethylauristatin F intended for research use as an ADC payload and microtubule-disrupting agent. The compound is supplied as a white to off-white solid, has molecular formula C39H58D8ClN5O8 and molecular weight 776.47, and is unstable in solution - prepare fresh solutions. Store sealed at 4°C and protect from moisture.
- High purity (99.7% by LCMS).
- High isotopic enrichment (99.8%).
- Soluble in DMSO at 100 mg/mL (may require ultrasonic).
- White to off-white solid appearance.
- Suitable as an antibody-drug conjugate (ADC) payload and microtubule-disrupting research reagent.
- Store sealed at 4°C and protect from moisture.
- Unstable in solution; prepare fresh solutions prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.