Learn More
Medchemexpress LLC 2-Pyrimidinamine, 4-[4-(1H-imidazol-1-yl)phenyl]-N-2-propyn-1-yl- | 2234272-10-5 | 98.1% | 50 MG
Supplier: Medchemexpress LLC HY13053050MG
AP-C5 is a selective inhibitor of guanosine 3′,5′-cyclic monophosphate (cGMP)-dependent protein kinase II (cGKII) with a pIC50 of 7.2. It is used for the research of diarrheal disease. This compound is also a click chemistry reagent containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
- Potent inhibition of cGMP-dependent cGKII-mediated protein phosphorylation
- Effective inhibition of cGMP-dependent, CFTR-mediated anion secretion in intestinal tissue
- Potentiates cAMP signaling through PDE inhibition
- Partially blocks heat-stable toxin (STa)-mediated short-circuit current (Isc) response in mouse ileum
- Click chemistry reagent with an alkyne group
- Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)