Learn More
Medchemexpress LLC Resazurin sodium | 62758-13-8 | 251.17 | 50 MG
Supplier: Medchemexpress LLC HY111391A1G
Resazurin sodium (Diazoresorcinol sodium) is a non-toxic, stable, and membrane-permeable blue non-fluorescent dye that is faintly fluorescent. It functions as a redox indicator, capable of being reduced in living cells to produce pink, highly fluorescent Resorufin (with excitation at 530-560 nm and emission at 590 nm). This makes Resazurin sodium useful for various cellular assays.
- Non-toxic and stable
- Membrane-permeable
- Functions as a redox indicator
- Can be reduced to highly fluorescent Resorufin in living cells (Ex=530-560 nm, Em=590 nm)
- Used for detecting cell viability
- Used for assessing toxicity
- Used for studying cell proliferation
- Used for investigating cell migration and invasion in various cell types (human, plant, animal, bacterial, and fungal)