Learn More
Medchemexpress LLC Resazurin (sodium) | 62758-13-8 | MFCD00005036 | 98.1% | 251.17 | 250MG
SDP
Supplier:  Medchemexpress LLC 250MGHY111391R
Encompass_Preferred
Resazurin sodium is a non-toxic, stable, membrane-permeable blue non-fluorescent dye that serves as a redox indicator. It is reduced to pink, highly fluorescent Resorufin in living cells, enabling various biological assays.
  • Used for detection of cell viability, toxicity, proliferation, migration, and invasion
  • Suitable for human, plant, animal, bacterial, and fungal cells
  • Stable and non-toxic
  • Membrane-permeable
Catalog No. 50-003-47546