Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-CO-C9-acid | 2172819-78-0 | 95.0% | 628.82 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY13934550MG

Encompass_Preferred

(S,R,S)-AHPC-CO-C9-acid is an E3 ligase ligand-linker conjugate that can be connected to the ligand for protein to form PROTACs. This compound is intended for research use only.

  • Functions as an E3 ligase ligand-linker conjugate.
  • Can be connected to ligand for protein to form PROTACs.
  • Exhibits antiproliferative activity against human NCI-H69 cells.
  • Has an IC50 > 40 μM against NCI-H69 cells.
  • Appearance: solid, white to off-white.
  • Ships at room temperature in continental US.
  • Storage conditions: 4°C, protect from light, stored under nitrogen.
  • Soluble in various solvents including DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.

Catalog No. 50-003-51322


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.