Learn More
Medchemexpress LLC FK 3311 | 116686-15-8 | MFCD00876455 | 98.7% | C15H13F2NO4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1444550MG
Encompass_Preferred
FK 3311, also known as COX-2 Inhibitor V, is a selective inhibitor of COX-2 with anti-inflammatory properties. It is intended for research use only.
  • Targets COX-2
  • Exhibits anti-inflammatory activity by inhibiting adjuvant-induced arthritis
  • Demonstrates analgesic effects in the acetic acid-induced writhing assay
  • Improves L-PVR, CO, PaO(2), and WDR in a canine model of pulmonary ischemia-reperfusion injury
  • Reduces histological tissue edema and PMN infiltration
  • Lowers serum TxB(2) levels and improves survival rate in pulmonary ischemia-reperfusion injury
  • Improves survival rates and lowers serum GOT/GPT levels in liver transplantation with ischemia-reperfusion injury
Catalog No. 50-003-54190