Learn More
Medchemexpress LLC Allyl (2-aminoethyl)carbamate | 223741-66-0 | 98.0% | C6H12N2O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY145735100MG
Encompass_Preferred
Allyl (2-aminoethyl)carbamate is identified as a cleavable linker, intended for research use only. This product appears as a colorless to light yellow liquid with a density of 1.039 g/cm³.
  • Not sold to patients
  • Suitable for Antibody-drug Conjugate/ADC related research
  • Store at 4°C in sealed storage, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-56245