Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antibacterial agent 125 | 1274611-43-6 | C15H11ClN2O | 1 ML
SDP

Supplier:  Medchemexpress LLC HY15191810MM/1ML

Encompass_Preferred

Antibacterial agent 125 is an antibacterial agent exhibiting potent antimicrobial activity against clinically relevant Gram-positive pathogens (MIC50 values 0.25-8 μM). It is suitable for antimicrobial resistance research.

  • Exhibits potent antimicrobial activity against Gram-positive pathogens.
  • Inhibits methicillin-resistant Staphylococcus aureus without cytotoxicity.
  • High metabolic stability and excellent safety profile.
  • Does not induce resistance after ten passages.
  • Moderately inhibits biofilm formation.
  • White to off-white solid appearance.
  • Purity of 98.90%.
  • Soluble in DMSO at 250 mg/mL (923.50 mM).
  • Molecular weight: 270.71.

Catalog No. 50-003-72327


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.