Learn More
Medchemexpress LLC Lamivudine | 134678-17-4 | 99.9% | 5 MG
Supplier: Medchemexpress LLC HYB0250R5MG
Lamivudine is an analytical standard intended for research and analytical applications. It is an orally active nucleoside reverse transcriptase inhibitor (NRTI) that can inhibit HIV reverse transcriptase 1/2 and the reverse transcriptase of hepatitis B virus. The salicylate form can penetrate the CNS.
- Analytical standard for research and analytical applications
- Inhibits HIV reverse transcriptase 1/2
- Inhibits hepatitis B virus reverse transcriptase
- Salicylate form can penetrate the CNS
- Commonly used in HPLC, GC, and MS research experiments
- For research use only