Learn More
Medchemexpress LLC Cyclic adenosine monophosphate sodium | 37839-81-9 | 100.0% | 100 MG
Supplier: Medchemexpress LLC HYB1511A100MG
Cyclic AMP (Cyclic adenosine monophosphate) sodium, an adenosine triphosphate derivative, is an intracellular signaling molecule responsible for directing cellular responses to extracellular signals. It is an important second messenger in many biological processes. It modulates mediator generation, suppresses the expression of pro-inflammatory cytokines, including TNF-α and IL-12, and enhances the production of the anti-inflammatory cytokine IL-10.
- Acts as an intracellular signaling molecule.
- Directs cellular responses to extracellular signals.
- Is an important second messenger in many biological processes.
- Modulates mediator generation.
- Suppresses pro-inflammatory cytokines.
- Enhances anti-inflammatory cytokine production.