Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Telenzepine dihydrochloride | 147416-96-4 | 99.57% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYB1789A50MG

Encompass_Preferred

Telenzepine dihydrochloride is a selective and orally active muscarinic M1 receptor antagonist with a Ki of 0.94 nM. It inhibits gastric acid secretion and has antiulcer effects. It is for research use only and not sold to patients.

  • Molecular weight: 443.39
  • Formula: C19H24Cl2N4O2S
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)

Catalog No. 50-004-11370


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.