Learn More
Medchemexpress LLC VTP50469 fumarate | 2169919-29-1 | 99.8% | 804.93 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY114162A10MM/1ML
Encompass_Preferred
VTP50469 fumarate is a highly selective and orally active Menin-MLL interaction inhibitor. It demonstrates potent anti-leukemia activity by targeting MLL-rearranged and NPM1-mutant leukemia. This compound is intended for research use only.
  • Potently inhibits Menin-MLL interaction with a Ki of 104 pM.
  • Inhibits cell proliferation in MLL-rearranged and ALL cell lines.
  • Induces apoptosis in MLL-r B cell ALL cell lines.
  • Promotes differentiation in MLL-r AML cell lines.
  • Displaces Menin from protein complexes.
  • Effective in in vivo models of leukemia, improving survival.
Catalog No. 50-004-35648