Learn More
Medchemexpress LLC (S)-2-(Cyclohexane-1-carbonyl-1,2,2,3,3,4,4,5,5,6,6-d11)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one | 98.2% | 323.47 | 1MG
Supplier: Medchemexpress LLC HY126057AS1MG
(S)-Praziquantel-d11 is the deuterium labeled (S)-Praziquantel, a toxic enantiomer of Praziquantel. (S)-Praziquantel is ineffective against worms. This product is for research use only and has not been fully validated for medical applications.
- Consistent with structure based on 1H NMR spectrum
- White to off-white (Solid) appearance