Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyruvate Dehydrogenase E2 Antibody (YA912) | 50 UL
SDP

Supplier:  Medchemexpress LLC HYP8107050UL

Encompass_Preferred

Pyruvate Dehydrogenase E2 Antibody (YA912) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to Pyruvate Dehydrogenase E2. For research use only.

  • Host: Mouse
  • Clonality: Monoclonal
  • Species reactivity: Human, Mouse
  • Predicted band size: 69 kDa
  • Observed band size: 69 kDa
  • Immunogen: Purified recombinant human Pyruvate Dehydrogenase E2 protein fragments expressed in E.coli
  • Application and dilution ratio: WB (1:500-1:1000), ICC/IF (1:50-1:200), IP (1:20)
  • Sensitivity: Endogenous
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Isotype: IgG1
  • Appearance: Liquid
  • Formulation: Supplied in PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide, pH 7.3
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
  • Shipping: With blue ice
  • Subcellular localization: Mitochondrion matrix

Catalog No. 50-004-42631


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.